ContactUsLinkComponent
PARK7 Human Recombinant fused to N-terminal His-Tag produced in E.coli is a single, non-glycosylated polypeptide chain containing 22 5 amino acids and having a molecular mass of 24 kDa. 1 * 5 µG
Catalog # BIRBORB81255-5
Supplier:  Biorbyt
PARK7 Human Recombinant fused to N-terminal His-Tag produced in E.coli is a single, non-glycosylated polypeptide chain containing 22 5 amino acids and having a molecular mass of 24 kDa. 1 * 5 µG
Catalog # BIRBORB81255-5
Supplier:  Biorbyt