ARL1 Recombinant Human produced in E.coli is a single, non-glycosylated polypeptide containing 201 amino acids (1-181 a.a.) and hav ing a molecular mass of 22.5 kDa. The ARL1 is fused to 20 amino acid His-Tag at N-terminus and purified by p 1 * 5 µG
Catalog # BIRBORB80925-5
Supplier: Biorbyt