ContactUsLinkComponent
HIV-1 p24 recombinant strain IIIB- is 51 kDa non-glycosylated polypeptide chain, containing the HIV-1 p24 immunodominant regions. 1 * 500 µG
Catalog # BIRBORB81693-500
Supplier:  Biorbyt
HIV-1 p24 recombinant strain IIIB- is 51 kDa non-glycosylated polypeptide chain, containing the HIV-1 p24 immunodominant regions. 1 * 500 µG
Catalog # BIRBORB81693-500
Supplier:  Biorbyt