ContactUsLinkComponent
MITF Human Recombinant (aa 170-279) expressed in E.coli, shows 38 kDa band on SDS-PAGE(Including GST).The MITF is purified by propri etary chromatographic techniques. 1 * 10 µG
Catalog # BIRBORB81216-10
Supplier:  Biorbyt
MITF Human Recombinant (aa 170-279) expressed in E.coli, shows 38 kDa band on SDS-PAGE(Including GST).The MITF is purified by propri etary chromatographic techniques. 1 * 10 µG
Catalog # BIRBORB81216-10
Supplier:  Biorbyt