Recombinant Human Holo Lactoferrin produced in Plant is glycosylated polypeptide chain having an approximate molecular mass of 77-80 kDa.The Human Holo Lactotransferrin is purified by proprietary chromatographic techniques. 1 * 1 g
Catalog # BIRBORB81203-1
Supplier: Biorbyt
Frequently Bought Together