ContactUsLinkComponent
ADAM10 extracellular domain minus the signal peptide and pro-sequence Human Recombinant fused to N-terminal His-Tag produced in E.co li is a single, non-glycosylated polypeptide chain containing 459 amino acids (214-672) and having a molecul 1 * 2 µG
Catalog # BIRBORB80900-2
Supplier:  Biorbyt
ADAM10 extracellular domain minus the signal peptide and pro-sequence Human Recombinant fused to N-terminal His-Tag produced in E.co li is a single, non-glycosylated polypeptide chain containing 459 amino acids (214-672) and having a molecul 1 * 2 µG
Catalog # BIRBORB80900-2
Supplier:  Biorbyt