ContactUsLinkComponent
CKMBITII Human Recombinant produced in Pichia Pastoris is glycosylated polypeptide chain having an identical amino acid sequence com pared to the native enzyme, purified under non-denaturing conditions and reacts with polyclonal antibodies t 1 * 10 µG
Catalog # BIRBORB80891-10
Supplier:  Biorbyt
CKMBITII Human Recombinant produced in Pichia Pastoris is glycosylated polypeptide chain having an identical amino acid sequence com pared to the native enzyme, purified under non-denaturing conditions and reacts with polyclonal antibodies t 1 * 10 µG
Catalog # BIRBORB80891-10
Supplier:  Biorbyt