Recombinant Lamin produced in E.coli is a single, non-glycosylated polypeptide chain containing 645 amino acids and having a molecul ar mass of 70 kDa. Lamin-A is major component of the nuclear lamina, dynamic meshwork located just under the 1 * 100 µG
Catalog # BIRBORB81193-100
Supplier: Biorbyt