ContactUsLinkComponent
LAIR1 Recombinant produced in E.coli is a single, non-glycosylated polypeptide chain containing amino acids 105 and having a molecul ar mass of 11.5 kDa. The extracellular domain of LAIR-1 (22-125aa) was overexpressed in E.coli, and purified 1 * 20 µG
Catalog # BIRBORB81191-20
Supplier:  Biorbyt
LAIR1 Recombinant produced in E.coli is a single, non-glycosylated polypeptide chain containing amino acids 105 and having a molecul ar mass of 11.5 kDa. The extracellular domain of LAIR-1 (22-125aa) was overexpressed in E.coli, and purified 1 * 20 µG
Catalog # BIRBORB81191-20
Supplier:  Biorbyt