ContactUsLinkComponent
Cytokeratin 20 Human Recombinant produced in E.coli is a single,non-glycosylated polypeptide chain having a molecular mass of 48,553 Dalton. The KRT20 is purified by proprietary chromatographic techniques. 1 * 20 µG
Catalog # BIRBORB81186-20
Supplier:  Biorbyt
Cytokeratin 20 Human Recombinant produced in E.coli is a single,non-glycosylated polypeptide chain having a molecular mass of 48,553 Dalton. The KRT20 is purified by proprietary chromatographic techniques. 1 * 20 µG
Catalog # BIRBORB81186-20
Supplier:  Biorbyt