ContactUsLinkComponent
Cytokeratin 19 Human Recombinant produced in E.coli is a single, non-glycosylated polypeptide chain having a molecular mass of 44,09 8 Dalton. The KRT19 is purified by proprietary chromatographic techniques. 1 * 20 µG
Catalog # BIRBORB81185-20
Supplier:  Biorbyt
Cytokeratin 19 Human Recombinant produced in E.coli is a single, non-glycosylated polypeptide chain having a molecular mass of 44,09 8 Dalton. The KRT19 is purified by proprietary chromatographic techniques. 1 * 20 µG
Catalog # BIRBORB81185-20
Supplier:  Biorbyt