KRT18 Human Recombinant produced in E.coli is a single, non-glycosylated polypeptide chain having calculated molecular mass of 51.6 kDa.The KRT18 is fused to His Tag at N-terminus and purified by proprietary chromatographic techniques. 1 * 10 µG
Catalog # BIRBORB81184-10
Supplier: Biorbyt