Cytokeratin 18 Human Recombinant produced in E.coli is a single, non-glycosylated polypeptide chain having calculated molecular mass of 48,201 Dalton, showing 45kDa band on SDS-page, pI-5.7.The KRT18 is purified by proprietary chromatograph 1 * 20 µG
Catalog # BIRBORB81183-20
Supplier: Biorbyt