KISS1 Human Recombinant produced in E.coli is a single, non-glycosylated polypeptide chain containing 120 amino acids (20-138) and h aving a molecular mass of 12.6 Dalton. The KISS1 is purified by proprietary chromatographic techniques. 1 * 2 µG
Catalog # BIRBORB81180-2
Supplier: Biorbyt