ContactUsLinkComponent
Recombinant KIR3DL1 produced in E.coli is a single, non-glycosylated polypeptide chain containing 132 amino acids and having a molec ular mass of 15 kDa.The KIR3DL1 is purified by proprietary chromatographic techniques. 1 * 20 µG
  BIRBORB81179-20
 :  Biorbyt
Recombinant KIR3DL1 produced in E.coli is a single, non-glycosylated polypeptide chain containing 132 amino acids and having a molec ular mass of 15 kDa.The KIR3DL1 is purified by proprietary chromatographic techniques. 1 * 20 µG
  BIRBORB81179-20
 :  Biorbyt