ContactUsLinkComponent
Recombinant KIR2DS4 produced in E.coli is a single, non-glycosylated polypeptide chain containing amino acids 1-202 and having a mol ecular mass of 22.2 kDa. The KIR2DS4 is purified by proprietary chromatographic techniques. 1 * 20 µG
Catalog # BIRBORB81178-20
Supplier:  Biorbyt
Recombinant KIR2DS4 produced in E.coli is a single, non-glycosylated polypeptide chain containing amino acids 1-202 and having a mol ecular mass of 22.2 kDa. The KIR2DS4 is purified by proprietary chromatographic techniques. 1 * 20 µG
Catalog # BIRBORB81178-20
Supplier:  Biorbyt