ContactUsLinkComponent
Recombinant KIR2DL3 produced in E.coli is a single, non-glycosylated polypeptide chain containing amino acids 1-202 and having a mol ecular mass of 22 kDa. The KIR2DL3 is purified by proprietary chromatographic techniques. 1 * 5 µG
Catalog # BIRBORB81177-5
Supplier:  Biorbyt
Recombinant KIR2DL3 produced in E.coli is a single, non-glycosylated polypeptide chain containing amino acids 1-202 and having a mol ecular mass of 22 kDa. The KIR2DL3 is purified by proprietary chromatographic techniques. 1 * 5 µG
Catalog # BIRBORB81177-5
Supplier:  Biorbyt