ContactUsLinkComponent
KIR2DL1 Recombinant produced in E.coli is a single, non-glycosylated polypeptide chain containing amino acids 1-202 and having a mol ecular mass of 22.2 kDa.The KIR2DL1 is purified by proprietary chromatographic techniques. 1 * 5 µG
Catalog # BIRBORB81176-5
Supplier:  Biorbyt
KIR2DL1 Recombinant produced in E.coli is a single, non-glycosylated polypeptide chain containing amino acids 1-202 and having a mol ecular mass of 22.2 kDa.The KIR2DL1 is purified by proprietary chromatographic techniques. 1 * 5 µG
Catalog # BIRBORB81176-5
Supplier:  Biorbyt