ContactUsLinkComponent
IPP-POZ Human Recombinant produced in E.coli is a single, non-glycosylated polypeptide chain containing 157 amino acids having a mol ecular mass of 17.3 kDa. 1 * 50 µG
Catalog # BIRBORB81173-50
Supplier:  Biorbyt
IPP-POZ Human Recombinant produced in E.coli is a single, non-glycosylated polypeptide chain containing 157 amino acids having a mol ecular mass of 17.3 kDa. 1 * 50 µG
Catalog # BIRBORB81173-50
Supplier:  Biorbyt