ContactUsLinkComponent
Involucrin Human Recombinant (aa 420-578) expressed in E.coli, shows 41 kDa band on SDS-PAGE (Icluding GST tag).The Involucrin is pu rified by proprietary chromatographic techniques. 1 * 10 µG
Catalog # BIRBORB81172-10
Supplier:  Biorbyt
Involucrin Human Recombinant (aa 420-578) expressed in E.coli, shows 41 kDa band on SDS-PAGE (Icluding GST tag).The Involucrin is pu rified by proprietary chromatographic techniques. 1 * 10 µG
Catalog # BIRBORB81172-10
Supplier:  Biorbyt