ContactUsLinkComponent
Human Serum Albumin contains 584 amino acid residues derived from the prototypical human serum albumin sequence. Suitable for use in biochemical, excipient (an inert substance used as diluent or vehicle for drug), culture media and chromato 1 * 100 mg
Catalog # BIRBORB81168-100
Supplier:  Biorbyt
Human Serum Albumin contains 584 amino acid residues derived from the prototypical human serum albumin sequence. Suitable for use in biochemical, excipient (an inert substance used as diluent or vehicle for drug), culture media and chromato 1 * 100 mg
Catalog # BIRBORB81168-100
Supplier:  Biorbyt