GNLY Human Recombinant produced in E.coli is a single, non-glycosylated, polypeptide chain containing double His Tag and having tota l molecular mass of 18.1 kDa. The GNLY is purified by proprietary chromatographic techniques. 1 * 10 µG
Catalog # BIRBORB81145-10
Supplier: Biorbyt