ContactUsLinkComponent
GLC8 Recombinant produced in Yeast is a single, glycosylated polypeptide chain containing 265 amino acids (1-229) and having a molec ular mass of 30.7 kDa. The GLC8 is fused to 35 amino acid His Tag at N-Terminus and purified by standard chr 1 * 50 µG
Catalog # BIRBORB81142-50
Supplier:  Biorbyt
GLC8 Recombinant produced in Yeast is a single, glycosylated polypeptide chain containing 265 amino acids (1-229) and having a molec ular mass of 30.7 kDa. The GLC8 is fused to 35 amino acid His Tag at N-Terminus and purified by standard chr 1 * 50 µG
Catalog # BIRBORB81142-50
Supplier:  Biorbyt