ContactUsLinkComponent
GADD45G Human Recombinant produced in E.coli is a single, non-glycosylated polypeptide chain containing 159 amino acids and having a molecular mass of 17.1 kDa. 1 * 10 µG
Catalog # BIRBORB81134-10
Supplier:  Biorbyt
GADD45G Human Recombinant produced in E.coli is a single, non-glycosylated polypeptide chain containing 159 amino acids and having a molecular mass of 17.1 kDa. 1 * 10 µG
Catalog # BIRBORB81134-10
Supplier:  Biorbyt