ContactUsLinkComponent
Recombinant Full-Length H3N2 A/Wyoming/2003/3 is glycosylated with N-linked sugars, produced using baculovirus vectors in insect cel ls and its Mw is 70,000 dalton. 1 * 10 µG
Catalog # BIRBORB81783-10
Supplier:  Biorbyt
Recombinant Full-Length H3N2 A/Wyoming/2003/3 is glycosylated with N-linked sugars, produced using baculovirus vectors in insect cel ls and its Mw is 70,000 dalton. 1 * 10 µG
Catalog # BIRBORB81783-10
Supplier:  Biorbyt