ContactUsLinkComponent
FOS-B Human Recombinant (a.a. 113-151) expressed in E.coli, shows 32 kDa SDS-PAGE (Including GST).The FOS-B is purified by proprieta ry chromatographic techniques. 1 * 5 µG
Catalog # BIRBORB81125-5
Supplier:  Biorbyt
FOS-B Human Recombinant (a.a. 113-151) expressed in E.coli, shows 32 kDa SDS-PAGE (Including GST).The FOS-B is purified by proprieta ry chromatographic techniques. 1 * 5 µG
Catalog # BIRBORB81125-5
Supplier:  Biorbyt