ContactUsLinkComponent
Factor VIIa Human Recombinant produced in BHK is glycosylated polypeptide two-chain dimer consisting of 406 amino acids with molecul ar weight of 50kD.The Factor-VIIa is purified by proprietary chromatographic techniques. 1 * 50 µG
Catalog # BIRBORB81109-50
Supplier:  Biorbyt
Factor VIIa Human Recombinant produced in BHK is glycosylated polypeptide two-chain dimer consisting of 406 amino acids with molecul ar weight of 50kD.The Factor-VIIa is purified by proprietary chromatographic techniques. 1 * 50 µG
Catalog # BIRBORB81109-50
Supplier:  Biorbyt