VAMP2 Human Recombinant produced in E.coli is a single, non-glycosylated polypeptide chain containing 126 amino acids (1-89) and hav ing a molecular mass of 13.8 kDa. The VAMP contains 37 amino acids His-Tag fused at N-terminus and purified 1 * 5 µG
Catalog # BIRBORB81460-5
Supplier: Biorbyt