ContactUsLinkComponent
Recombinant human ubiquitin featuring Lys 48 to Arg48 mutation is useful for the reduction of poly-Ub chain length and conjugation r ates.Ubiquitin K48R is expressed in E.coliand purified by proprietary chromatographic techniques. 1 * 20 µG
Catalog # BIRBORB81454-20
Supplier:  Biorbyt
Recombinant human ubiquitin featuring Lys 48 to Arg48 mutation is useful for the reduction of poly-Ub chain length and conjugation r ates.Ubiquitin K48R is expressed in E.coliand purified by proprietary chromatographic techniques. 1 * 20 µG
Catalog # BIRBORB81454-20
Supplier:  Biorbyt