ContactUsLinkComponent
FABP5 Human Recombinant produced in E.coli is a single, non-glycosylated polypeptide chain containing 135 amino acids and having a m olecular mass of 19.66kDa. FABP5 is fused to His tag at N-terminus and purified by standard chromatography t 1 * 25 µG
Catalog # BIRBORB81103-25
Supplier:  Biorbyt
FABP5 Human Recombinant produced in E.coli is a single, non-glycosylated polypeptide chain containing 135 amino acids and having a m olecular mass of 19.66kDa. FABP5 is fused to His tag at N-terminus and purified by standard chromatography t 1 * 25 µG
Catalog # BIRBORB81103-25
Supplier:  Biorbyt