ContactUsLinkComponent
FABP3 Human Recombinant produced in E.coli is a single, non-glycosylated polypeptide chain containing 133 amino acids and having a m olecular mass of 19.1 kDa. FABP3 is fused to 37 amino acid His tag at N-terminus and purified by standard ch 1 * 5 µG
Catalog # BIRBORB81099-5
Supplier:  Biorbyt
FABP3 Human Recombinant produced in E.coli is a single, non-glycosylated polypeptide chain containing 133 amino acids and having a m olecular mass of 19.1 kDa. FABP3 is fused to 37 amino acid His tag at N-terminus and purified by standard ch 1 * 5 µG
Catalog # BIRBORB81099-5
Supplier:  Biorbyt