ContactUsLinkComponent
ETS1 Human Recombinant (aa 220-442) expressed in E.coli, shows 48 kDa band on SDS-PAGE (Including GST).The ETS1 is purified by propr ietary chromatographic techniques. 1 * 2 µG
Catalog # BIRBORB81095-2
Supplier:  Biorbyt
ETS1 Human Recombinant (aa 220-442) expressed in E.coli, shows 48 kDa band on SDS-PAGE (Including GST).The ETS1 is purified by propr ietary chromatographic techniques. 1 * 2 µG
Catalog # BIRBORB81095-2
Supplier:  Biorbyt