ContactUsLinkComponent
ERP44 Recombinant Human produced in E.coli is a single, non-glycosylated polypeptide chain containing 415 amino acids (30-406 a.a.) and having a molecular mass of 48 kDa. The ERP44 is fused to 38 amino acid His-Tag at N-terminus and purifie 1 * 25 µG
Catalog # BIRBORB81093-25
Supplier:  Biorbyt
ERP44 Recombinant Human produced in E.coli is a single, non-glycosylated polypeptide chain containing 415 amino acids (30-406 a.a.) and having a molecular mass of 48 kDa. The ERP44 is fused to 38 amino acid His-Tag at N-terminus and purifie 1 * 25 µG
Catalog # BIRBORB81093-25
Supplier:  Biorbyt