ContactUsLinkComponent
Ephrin B3 Human Recombinant (a.a. 135-341) expressed in E.coli, shows 47 kDa SDS-PAGE.The Ephrin B3 is purified by proprietary chrom atographic techniques. 1 * 2 µG
Catalog # BIRBORB81090-2
Supplier:  Biorbyt
Ephrin B3 Human Recombinant (a.a. 135-341) expressed in E.coli, shows 47 kDa SDS-PAGE.The Ephrin B3 is purified by proprietary chrom atographic techniques. 1 * 2 µG
Catalog # BIRBORB81090-2
Supplier:  Biorbyt