ContactUsLinkComponent
Human Cardiac Troponin produced from Human Cardiac Tissue, having molecular weight of approximately 35 kDa.The Cardiac troponin-T is purified using combination of ion-exchange and affinity chromatography steps. 1 * 50 µG
Catalog # BIRBORB81438-50
Supplier:  Biorbyt
Human Cardiac Troponin produced from Human Cardiac Tissue, having molecular weight of approximately 35 kDa.The Cardiac troponin-T is purified using combination of ion-exchange and affinity chromatography steps. 1 * 50 µG
Catalog # BIRBORB81438-50
Supplier:  Biorbyt