ContactUsLinkComponent
Recombinant Human TNI produced in E.coli is a single, non-glycosylated, polypeptide chain containing 165 amino acids and having a mo lecular mass of 24,016 Dalton.The TNI is purified by proprietary chromatographic techniques. 1 * 50 µG
Catalog # BIRBORB81437-50
Supplier:  Biorbyt
Recombinant Human TNI produced in E.coli is a single, non-glycosylated, polypeptide chain containing 165 amino acids and having a mo lecular mass of 24,016 Dalton.The TNI is purified by proprietary chromatographic techniques. 1 * 50 µG
Catalog # BIRBORB81437-50
Supplier:  Biorbyt