TPM1 Human Recombinant produced in E.coli is signle, non-glycosylated, polypeptide chain containing 304 amino acids (1-284 a.a.)and having total molecular mass of 35 kDa. TPM1 is fused to 20 amino acid His Tag at N-terminus and is purified 1 * 2 µG
Catalog # BIRBORB81425-2
Supplier: Biorbyt