ContactUsLinkComponent
EIF2S1 Recombinant Human produced in E.coli is a single, non-glycosylated polypeptide chain containing 335 amino acids (1-315 a.a.) and having a molecular mass of 38.2 kDa. The EIF2S1 is fused to 20 amino acid His-Tag at N-terminus and puri 1 * 10 µG
Catalog # BIRBORB81080-10
Supplier:  Biorbyt
EIF2S1 Recombinant Human produced in E.coli is a single, non-glycosylated polypeptide chain containing 335 amino acids (1-315 a.a.) and having a molecular mass of 38.2 kDa. The EIF2S1 is fused to 20 amino acid His-Tag at N-terminus and puri 1 * 10 µG
Catalog # BIRBORB81080-10
Supplier:  Biorbyt