Endothelial Mitogen can be used for Human Mammalian Vascular Endothelial Cells at concentration of 0.1-0.3mg/ml of serum supplemente d media 0.01-0.1mg with heparin. Endothelial Mitogen can be used for Hybridoma Cell Cloning at concentration 1 * 20 mg
Catalog # BIRBORB81077-20
Supplier: Biorbyt