ESAT-6 Recombinant produced in E.coli is a single, non-glycosylated, polypeptide having total molecular mass of 11261.35 Dalton.The ESAT-6 contains additional amino acid residues His-Tag. 1 * 10 µG
Catalog # BIRBORB81075-10
Supplier: Biorbyt