DYNLRB1 Recombinant Human produced in E.coli is a single, non-glycosylated polypeptide chain containing 104 amino acids (1-96 a.a.) and having a molecular mass of 11.9 kDa. The DYNLRB1 is fused to amino acid His-Tag at C-terminus and purifi 1 * 2 µG
BIRBORB81074-2
: Biorbyt