DR1 Recombinant Human produced in E.coli is a single, non-glycosylated polypeptide chain containing 196 amino acids (1-176 a.a.) and having a molecular mass of 21.6 kDa. The DR1 is fused to 20 amino acid His-Tag at N-terminus and purified b 1 * 25 µG
Catalog # BIRBORB81072-25
Supplier: Biorbyt