Recombinant Human THBS1 is glycosylated with N-linked sugars and produced using baculovirus vectors in insect cells. Recombinant Hum an THBS1 is Reactive with A4.1 anti-TSP mAb and its Mw is 140,000 Dalton. 1 * 10 µG
Catalog # BIRBORB81417-10
Supplier: Biorbyt