Syndecan-4 Human Recombinant fused with 6X His tag produced in E.coli is a single, non-glycosylated, polypeptide chain containing 14 0 amino acids and having a molecular mass of 15.4 kDa. The SDC4 is purified by proprietary chromatographic t 1 * 50 µG
Catalog # BIRBORB81404-50
Supplier: Biorbyt