SUMO2 Human Recombinant produced in E.coli is a single, non-glycosylated polypeptide chain containing 93 amino acids and having a mo lecular mass of 10.6 kDa.The SUMO-2 is purified by proprietary chromatographic techniques. 1 * 10 µG
Catalog # BIRBORB81400-10
Supplier: Biorbyt