Recombinant Streptavidin-NC produced in E.coli is a single, non-glycosylated, polypeptide chain having a molecular mass of 24kDa. Re combinant Streptavidin-NC not only binds to nitrocellulose membrane readily but also preserves the full biot 1 * 20 µG
Catalog # BIRBORB81397-20
Supplier: Biorbyt