CSTB Human Recombinant fused to His-Tag at N-Terminus produced in E.coli is a single, non-glycosylated polypeptide chain containing 118 amino acids and having a molecular mass of 13 kDa. 1 * 20 µG
Catalog # BIRBORB81053-20
Supplier: Biorbyt