Recombinant Human CRP produced in E.coli is non-glycosylated polypeptide chain having total molecular mass of 115 kDa that correspon ds to the pentamer structure of 23 kDa monomer determined by amino acid sequence. The CRP is purified by pro 1 * 500 µG
Catalog # BIRBORB81051-500
Supplier: Biorbyt