ContactUsLinkComponent
CRADD Human Recombinant produced in E.coli is a single, non-glycosylated polypeptide chain containing 219 amino acids (1-199) and ha ving a molecular mass of 24.9 kDa. CRADD is fused to 20 amino acids His-Tag at N-terminus. 1 * 5 µG
Catalog # BIRBORB81048-5
Supplier:  Biorbyt
CRADD Human Recombinant produced in E.coli is a single, non-glycosylated polypeptide chain containing 219 amino acids (1-199) and ha ving a molecular mass of 24.9 kDa. CRADD is fused to 20 amino acids His-Tag at N-terminus. 1 * 5 µG
Catalog # BIRBORB81048-5
Supplier:  Biorbyt