ContactUsLinkComponent
Recombinant SIV p55- Strains: SIV mac 23g and SIV smH4 is glycosylated with N-linked sugars and produced using baculovirus vectors i n insect cells. 1 * 10 µG
  BIRBORB81740-10
 :  Biorbyt
Recombinant SIV p55- Strains: SIV mac 23g and SIV smH4 is glycosylated with N-linked sugars and produced using baculovirus vectors i n insect cells. 1 * 10 µG
  BIRBORB81740-10
 :  Biorbyt